Window Washing Tools, Micronova
Supplier: Micronova
Certificates
About this item
Tools feature a T-Mop™ on one side, and a squeegee on the other. Ideal for cleaning windows and other smooth surfaces.
- Simple and efficient method for cleaning windows to a streak-free finish
- Attaches to the STA series of fixed length and extendible plastic handles
- Available in 12 or 18" widths
- Covers available in NovaPoly™, PolySorb™, Microfiber, and MegaTex™ materials
- Covers available sterile
The Window Washing Tool is a simple and efficient method for cleaning windows and other smooth surfaces in clean and controlled environments. Dual surfaces allow the operator to apply detergent or rinsing agent with the mop surface, and then flip the unit to squeegee for a streak-free finish. The Window Washing Tool features an ACME thread and can be attached to the STAH or STAEH series of fixed length and extendible plastic handles. The entire unit is autoclavable and will hold up to strong disinfectants and detergents.
Frequently Bought Together