Normal Human Serum, GeneTex
Catalog # 89347-956
Supplier: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Sterility:Sterile
- Description:Normal Human Serum
- Size:100 ml
- Cat. no.:89347-956
- Supplier No.:GTX73261
Specifications
About this item
Normal human serum is manufactured from individual units of off clot serum (serum collected from spontaneously clotted whole blood).
- Liquid in form
- Store at –80 °C
Used in blocking, ELISA, ICC/IF, IHC and WB.
Caution: For In vitro laboratory use only. Not for any clinical, therapeutic, or diagnostic use in humans or animals. Not for animal or human consumption.