High Pressure Sample Pans, PerkinElmer
Fournisseur: PerkinElmer
Certificats
À propos de cet article
Pans contain the sample in a sealed environment throughout an experiment.
- Pressure ranges from 24 to 150 bar
The sealing suppresses the vaporization of a solvent or contains a volatile reaction product, thereby eliminating the interfering effects of the heat of vaporization.
The stainless steel pans with covers and seals are reusable and includes 5 pans and 20 gold-plated seals.
Frequently Bought Together