Anti-Static Tapes, Botron
Anti-Static Tapes, Botron
Anti-Static Tapes, Botron
Anti-Static Tapes, Botron
These clear anti static tapes are perfect for sealing ESD bags and ESD packing containers. They can also be used for a wide range of workstation applications such as binding, edging and securing.
Product Family Options
  • Désignation
    Color
    Core Size
    Longueur
    Adhesion
    Largeur
    Épaisseur
    Elongation
    Matière
    Tensile Strength
  • ESD cellulose tape, transparent
    Clear
    25.4 mm (1")
    108 ft.
    66 oz./in.
    ¾"
    2 mil.
    18%
    Transparent Cellulose
    90 N/25 mil
    Each
    Each - $8.35
    $8.35
    Each
       $8.35
     : B1631
  • ESD cellulose tape, transparent
    Clear
    76 mm (3")
    216 ft.
    66 oz./in.
    ¾"
    2 mil.
    18%
    Transparent Cellulose
    90 N/25 mil
    Each
    Each - $12.70
    $12.70
    Each
       $12.70
     : B1633
  • ESD cellulose tape, transparent
    Clear
    25.4 mm (1")
    108 ft.
    66 oz./in.
    ½"
    2 mil.
    18%
    Transparent Cellulose
    90 N/25 mil
    Each
    Each - $6.39
    $6.39
    Each
       $6.39
     : B1651
  • ESD cellulose tape, transparent
    Clear
    76 mm (3")
    216 ft.
    66 oz./in.
    ½"
    2 mil.
    18%
    Transparent Cellulose
    90 N/25 mil
    Each
    Each - $8.14
    $8.14
    Each
       $8.14
     : B1653
  • ESD cellulose tape, transparent
    Clear
    25.4 mm (1")
    108 ft.
    66 oz./in.
    1"
    2 mil.
    18%
    Transparent Cellulose
    90 N/25 mil
    Each
    Each - $10.79
    $10.79
    Each
       $10.79
     : B1611
  • ESD cellulose tape, transparent
    Clear
    76 mm (3")
    216 ft.
    66 oz./in.
    1"
    2 mil.
    18%
    Transparent Cellulose
    90 N/25 mil
    Each
    Each - $15.73
    $15.73
    Each
       $15.73
     : B1613
  • ESD cellulose tape, transparent
    Clear
    76 mm (3")
    216 ft.
    66 oz./in.
    2"
    2 mil.
    18%
    Transparent Cellulose
    90 N/25 mil
    Each
    Each - $30.45
    $30.45
    Each
       $30.45
     : B1623