ReLeSR™ | Avantor
ReLeSR™
ReLeSR™, 100 ml
Catalog # MSPP-05872
Marketsource
Fournisseur #:  100-0483
ReLeSR™
Catalog # MSPP-05872
Marketsource
ReLeSR™
ReLeSR™
ReLeSR™, 100 ml
Price
$130.77
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
cGMP, enzyme-free human pluripotent stem cell selection and passaging reagent.
  • Simplify cell passaging with a straightforward protocol
  • Eliminate the need for manual removal (selection) of differentiated cells
  • Generate cell aggregates without manual scraping
  • Passage cells in flasks and large culture vessels
  • Obtain quality cutures with this gentle, chemically defined, enzyme-free, GMP solution
  • Achieve high expansion of human ES/iPS cells after passaging
Efficiently dissociate and passage human embryonic stem (ES) or induced pluripotent stem (iPS) cells as aggregates without manual selection or scraping with ReLeSR™. Passaging human ES/iPS cells with ReLeSR™ easily generates optimally-sized aggregates, while eliminating the hassle and variability associated with manual manipulation. By eliminating the need for manual scraping, this enzyme-free reagent enables the use of culture flasks and other closed vessels, facilitating cell culture scale-up and automation. ReLeSR™ is manufactured following relevant cGMPs under a certified quality management system to ensure the highest quality and consistency for reproducible results.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Spécifications
  • Désignation:
    ReLeSR™
  • Taille:
    100 ml