XTT Cell Proliferation Assay Kits, Cayman Chemical
Fournisseur: Cayman Chemical Company
Certificats
À propos de cet article
A colorimetric, non-radioactive assay for assessing cell viability and proliferation. Tetrazolium salt, XTT, is reduced to a soluble purple formazan by trans-plasma membrane electron transport from NADH via an electron mediator.
- Detect 500 to 100000 cells per well
- Allows investigators to screen drug candidates involved in cell cycle regulation
Cell proliferation assays are widely used to assess cell cycle regulatory factors such as growth factors, cytokines, mitogens, and drugs.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together