Ward's Lab Wipe Holders | Avantor
Ward's Lab Wipe Holders
Supplier:   Heathrow Scientific
Ward's Lab Wipe Holders
Certificates
Ward's Lab Wipe Holders
Ward's Lab Wipe Holders
2 Options Available Below
Product Details & Documents
Holds lab wipes such as Kimwipes® and other major brands.
  • Mounts on a shelf or wall
  • Mounting materials included
Clear acrylic holders fit boxes of small or large lab wipes. The smaller version attaches with double-stick tape and the larger version with screws.
Product Family Options
Informations sur le produit
  • Désignation
    Dimensions
    Availability
    Price
  • Small Lab Wipe Holder
    118×122×84 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $87.85
    $87.85
    Each
       $87.85
    Supplier Number: HS234521W
  • Large Lab Wipe Holder
    320×127×97 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $114.85
    $114.85
    Each
       $114.85
    Supplier Number: HS234522W