Ward's Adapt-A-Rack | Avantor
Ward's Adapt-A-Rack
Supplier:   Heathrow Scientific
Ward's Adapt-A-Rack
Certificates
Ward's Adapt-A-Rack
Ward's Adapt-A-Rack
3 Options Available Below
Product Details & Documents
Flexible multi-tube rack.
  • Holds different sized tubes in each slot
  • Racks can be linked together for additional holding capacity
  • Open sides allow for label viewing
  • Flat base to hold tubes upright
  • Linked racks should be supported underneath when carrying
  • Available in color combinations for easy coding and identification
  • Autoclavable
Each slot on the Ward's Adapt-A-Rack adapts to accommodate 12 to 30 mm (5 to 50 ml) tube sizes and holds them in a firm upright position. Each rack is made from POM and contains four wells.
Product Family Options
Informations sur le produit
  • Color
    Dimensions
    Availability
    Price
  • Blue/Green and Green/Blue
    18.1×5.6×7.6 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $34.70
    $34.70
    Log in pour la disponibilité
    Case of 20
    Case of 20 - $346.55
    $346.55
    Pack of 2
       $34.70
    Case of 20
       $346.55
    Supplier Number: HS120184W
  • Blue/Yellow and Yellow/Blue
    18.1×5.6×7.6 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $34.70
    $34.70
    Log in pour la disponibilité
    Case of 20
    Case of 20 - $346.55
    $346.55
    Pack of 2
       $34.70
    Case of 20
       $346.55
    Supplier Number: HS120185W
  • Pink/White and White/Pink
    18.1×5.6×7.6 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $34.70
    $34.70
    Log in pour la disponibilité
    Case of 20
    Case of 20 - $346.55
    $346.55
    Pack of 2
       $34.70
    Case of 20
       $346.55
    Supplier Number: HS120186W