Ward's TracRack®
Ward's TracRack®
Supplier:   Heathrow Scientific
Ward's TracRack®
Certificates
Ward's TracRack®
Ward's TracRack®
1 Options Available Below
Product Details & Documents
Tube storage and a tube rack in one.
  • Helps reduce potential cross contamination
  • Saves valuable working space and allows more room for sample manipulation
  • Stops opened microfuge lids from interfering with other wells
  • Alphanumeric well identification
  • Tube segments sold with the box and lid
Removing and working with samples from cold temperature storage has never been easier than with the Ward's TracRack®. This rack and storage system eliminates the hassle of taking an entire rack of samples out of cold storage because the TracRack® enables lab professionals to remove one row of samples at a time.
Eight individual segments are nested in a 2" polypropylene film box that allows users to easily open, identify, and remove only the segments of samples needed. Once the segments are removed, simply replace the box lid and leave the remaining samples in storage. Each tube segment is made from durable polypropylene and allows for easy working while saving valuable counter space. Two versions are available.
Product Family Options
Product Information
  • Color
    Nb de places
    For Tubes
    Dimensions
    Availability
    Price
  • Purple
    64
    1.5/2.0 ml
    12.93×13×3.5 cm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $91.00
    $91.00
    Log in for availability
    Case of 10
    Case of 10 - $884.80
    $884.80
    Each
       $91.00
    Case of 10
       $884.80
  • 0.5/0.2 ml
    Discontinued