Ward's Benchtop Counter
Ward's Benchtop Counter
Supplier:   Heathrow Scientific
Ward's Benchtop Counter
Certificates
Ward's Benchtop Counter
Ward's Benchtop Counter
2 Options Available Below
Product Details & Documents
Manual counting made simple.
  • Each key records up to 999
  • Six and nine key options available
Ward's mechanical, benchtop counters are perfect for blood cells (label included), bacterial colonies, or other repetitive counting. Total window keeps track of the total number of strokes made on all other keys. A bell rings when total reaches 100 Blood cell key labels. 6-Key: baso., eosin., mono., lymph., poly., and total; 9-Key: baso., eosin., myelo., juven., stab., seg., lymph., mono., and total.
Product Family Options
Product Information
  • Désignation
    Dimensions
    Availability
    Price
  • 6-key Benchtop Counter
    25×7.6×5.6 cm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $714.20
    $714.20
    Log in for availability
    Case of 10
    Case of 10 - $7,142.00
    $7,142.00
    Each
       $714.20
    Case of 10
       $7,142.00
    Supplier Number: HS5360AW
  • 9-Key Benchtop Counter
    32×7.6×5.6 cm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $1,043.10
    $1,043.10
    Log in for availability
    Case of 10
    Case of 10 - $10,431.00
    $10,431.00
    Each
       $1,043.10
    Case of 10
       $10,431.00
    Supplier Number: HS5360BW