Human gamma Globulins Fraction II, MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Used as an immunological challenge in studies of mechanisms of immunotolerance.
Each donor unit used in the preparation of the product was tested and found negative for Anti-HIV 1/2, HBsAg, Anti-HCV and HIV-1 Ag by FDA approved methods or CE approved methods.
Specifications
- Species:Human
- Storage conditions:Store at 2-8 °C
- Protein/peptide name:gamma Globulins Fraction II
- Purity:Protein: 97.0% (By Biuret)