About this item
Albumins are a group of simple proteins found in the body fluids and tissues of animals, and in some plant seeds. It is a single polypeptide chain with one free sulfhydryl group (Cys34) and 17 intrachain disulfide bonds.
Prepared using Cohn method IV. The material is non-reactive for HbsAg, Anti-HCV 3.0, Anti-HIV-1/2 and STS (RPR). This material is also negative for HIV-1 and HCV by PCR
ALBUMIN, HUMAN is recommended for use in absorption, distribution, metabolism, and excretion (ADME) pharmacological research; cell culture; drug delivery research; and cryopreservation of cells.
Albumins are soluble monomeric proteins found in the body fluids and tissues of animals and in some plant seeds. Serum albumin functions as a carrier protein for steroids, fatty acids, and thyroid hormones. Serum albumins are also vital in regulating the colloidal osmotic pressures of blood.
Specifications
- Species:Human
- Storage conditions:Store at 2-8 °C
- Protein synonyms:HSA|rHSA
- Protein/peptide name:Albumin
- Purity:97-99%