RBC Lysis Buffer, IBI Scientific | Avantor
RBC Lysis Buffer, IBI Scientific
Supplier:   IBI Scientific
RBC Lysis Buffer, IBI Scientific
Supplier:  IBI Scientific
Certificates
RBC Lysis Buffer, IBI Scientific
3 Options Available Below
Product Details & Documents
RBC Lysis Buffer is used to lyse red blood cells in nucleic acid isolation applications.
  • Lyses red blood cells for DNA and RNA extractions
Can be used with the Genomic DNA (Blood) Extraction Kits and the Total RNA (Blood) Extraction Kits.
Product Family Options
Informations sur le produit
  • Taille
    Availability
    Price
  • 135 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $67.42
    $67.42
    Each
       $67.42
    Supplier Number: IB47620
  • 200 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $89.40
    $89.40
    Each
       $89.40
    Supplier Number: IB47621
  • 500 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $171.34
    $171.34
    Each
       $171.34
    Supplier Number: IB47622