MX200S Protective Eyewear, HexArmor
MX200S Protective Eyewear, HexArmor
Supplier:   HexArmor
MX200S Protective Eyewear, HexArmor
Supplier:  HexArmor
Certificates
MX200S Protective Eyewear, HexArmor
MX200S Protective Eyewear, HexArmor
4 Options Available Below
Product Details & Documents
Safety eyewear should never be treated as one size fits all. Simply wearing any generic pair of safety eyewear isn’t enough to protect the eyes.
  • Slim fit design for those who need a narrower fit
  • Rimless lens design with wide field of vision
  • Dual-injection soft-grip side arms for comfort and non-slip fit
  • Beveled lens edge for additional upper eye protection
  • Metal-free hinge design and integrated channel for eye ventilation
  • Contoured nose bridge for comfort and fit
  • LTX™ Lens Technology for premium optical clarity with 100% UV protection
The MX200S with permanent anti-fog coatings. Ideal for those who need a narrower fit, the MX200S (a close relative to the MX200), has a sharp, rimless, wraparound lens design that delivers maximum protection plus high levels of durability and comfort thanks to the unique dual-injection mold technology.
Although it may decrease the possibility of injury, poorly or incorrectly fitting eyewear will put users at a higher risk for injury. HexArmor’s German-engineered dual-injection mold technology, allows the combination of soft and hard components into flex frames, making them comfortable and durable, yet highly adaptive.
They fit to most face widths or shapes, making wearability, protection, and compliance an easier endeavor.
Certifications: Meets Standards ANSI Z87.1.2015, CE EN 166, CSA 94.3.
Ordering information: Attachable foam guard available.
Product Family Options
Informations sur le produit
  • Désignation
    Lens Coating
    Lens Tint
    Availability
    Price
  • Protective eyewear MX200s
    Anti-fog, scratch-resistant/hardcoat (TruShield®)
    Clear
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $28.41
    $28.41
    Each
       $28.41
    Supplier Number: 11-11001-02
  • Protective eyewear MX200s
    Anti-fog, scratch-resistant/hardcoat (TruShield® 2SF)
    Clear
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $33.51
    $33.51
    Each
       $33.51
    Supplier Number: 11-11003-05
  • Protective eyewear MX200s
    Anti-fog, scratch-resistant/hardcoat (TruShield®)
    Amber
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $30.82
    $30.82
    Each
       $30.82
    Supplier Number: 11-11004-02
  • Protective eyewear MX200s
    Anti-fog, scratch-resistant/hardcoat (TruShield®)
    Gray, 23%
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $29.87
    $29.87
    Each
       $29.87
    Supplier Number: 11-11005-02
  • Protective eyewear MX200s
    Anti-fog
    Sliver mirror, 53%
    Discontinued
    Supplier Number: -