LaMotte® Turbidity Test Kit | Avantor
LaMotte® Turbidity Test Kit
Turbidity Test Kit, 1 Kit, 100
Catalog # 470227-856
Supplier:   LaMotte
Fournisseur #:  5887
LaMotte® Turbidity Test Kit
Catalog # 470227-856
Supplier:  LaMotte
CAS Number:  
LaMotte® Turbidity Test Kit
LaMotte® Turbidity Test Kit
Turbidity Test Kit, 1 Kit, 100
CAS Number:  
Price
$77.80
Log in pour la disponibilité
Each
Product Details & Documents
This kit tests for turbidity using a turbidmetric card used match clarity to your sample.
  • Unlimited tests
Range and sensitivity is 0, 20, 40, 60, 80, 100 JTU.
Spécifications
Spécifications
  • Désignation:
    Turbidity Test Kit
  • Taille:
    1 Kit
  • No. of Tests:
    100
Spécifications