Human beta-2-Microglobulin (from Urine), MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Lyophilized from 0.02M Ammonium Bicarbonate (may contain traces of buffer salt).
Immunological identity verified by radial immunodiffusion. Cellulose acetate electrophoresis shows one band indicating β2 electrophoretic mobility. SDS-PAGE shows one band corresponding to a molecular weight of approximately 12,000.
Reconstitution: To reconstitute use a phosphate buffer, pH >7.0, containing 0.15 M NaCl.
- Calibrators in enzyme assays
- Cancer and renal biomarker
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Source:Urine
- Species:Human
- Storage conditions:Store at 2-8 °C. For long-term storage after reconstitution, aliquot and store at -20 °C.
- Biological activity:Bioburden: <100 CFU/mL, Biohazard: Raw materials tested negative to HBsAg, HIV 1/2 antibodies, Hepatitis C antibodies and Syphilis by FDA approved methods. Since no test can offer complete assurance handle with precaution.
- Protein/peptide name:beta-2-Microglobulin
- Purity:>98% by SDS PAGE and Cellulose Acetate Electrophoresis.
- Formulation:Lyophilized powder
- Grade:Iodination Grade
- Tested applications:Immunoassays