Order Entry
Canada
ContactUsLinkComponent
Human IgM, MP Biomedicals
Human IgM, MP Biomedicals
Catalog # IC0855916
Supplier:  MP Biomedicals
CAS Number:  
undefined
Human IgM, MP Biomedicals
Catalog # IC0855916
Supplier:  MP Biomedicals
Supplier Number:  0855916
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Source:
    Human
  • Species:
    Human
  • Size:
    5 mg
  • Storage conditions:
    Store unopened product at 2-8 °C. Store opened material no longer than 1-2 days at 2-8 °C unless sodium azide is added. Do not freeze.
  • Protein/peptide name:
    IgM
  • Purity:
    >99%, liquid
  • Concentration:
    Total Protein: 5.3 mg/mL; Volume: 19.4 mL
  • Tested applications:
    Immunoassays
  • Cat. no.:
    IC0855916

Specifications

About this item

Product is human IgM and buffer salts. From human myeloma, highly purified, immunoabsorbed to remove trace amounts of contaminant proteins.

  • Standard in ELISA (100 μg/ml)
  • Dot blot assay
  • Purified human IgM is suitable for use as a positive control in antibody identification screening techniques and is recommended as an ELISA coating antigen and control.
  • From human myeloma, highly purified, immunoabsorbed to remove trace amounts of contaminant proteins.

The product is tested for purity at a minimum of 20 mg/mL using immunoelectrophoresis. The product is human IgM; no trace of IgG, IgA or non-immunoglobulin serum proteins is detected. The total protein is determined using the Biuret procedure, with bovine albumin as the standard. Each vial is overfilled by approximately 10 percent so that a minimum of 5 mg of human IgM is recovered.

Human IgM is purified from human myeloma serum using multi-step procedures which may include salt fractionation, gel filtration, ion-exchange chromatography and immunoabsorption. The product is dialyzed into 0.01M sodium phosphate, 0.07M sodium chloride, pH 7.3, filtered through a 0.22 μm filter and vialed. No preservative has been added.