Order Entry
Canada
ContactUsLinkComponent
Mouse Albumin Fraction V, MP Biomedicals
Catalogue # IC0855858
Fournisseur:  MP Biomedicals
Numéro CAS:  
undefined
Mouse Albumin Fraction V, MP Biomedicals
Catalogue # IC0855858
Fournisseur:  MP Biomedicals
Supplier Number:  0855858
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Spécifications

  • Species:
    Mouse
  • Size:
    50 mg
  • Storage conditions:
    Store unopened product at 2-8 °C. Store reconstituted material no longer than a few days at 2-8 °C unless sodium azide is added. For long-term storage, aliquot and freeze at –20 °C.
  • Reconstitution instructions:
    Reconstitute product with 3.0 mL of deionized or distilled water. Gentle swirling may be used to speed rehydration. Avoid vigorous shaking of the reconstituted material.
  • Protein/peptide name:
    Albumin Fraction V
  • Concentration:
    Total Protein: 16.8 mg/mL; Volume: 3.0 mL
  • Formulation:
    Lyophilized
  • Tested applications:
    Immunoassays
  • Cat. no.:
    IC0855858

Spécifications

À propos de cet article

Product is the lyophilized powder of mouse albumin Fraction V and buffer salts.

  • Albumin, Mouse can be used for making immune complexes with anti-albumin antibodies.
  • Fatty acids and globulins will not interfere with the complex formation.

The total protein is determined using the Biuret procedure, with bovine albumin as the standard. Each vial is overfilled by approximately 10 percent so that a minimum of 50 mg of mouse albumin Fraction V is obtained upon reconstitution.

Mouse albumin Fraction V is purified from pooled mouse serum using multi-step procedures which may include salt fractionation, gel filtration and ion-exchange chromatography. The product is dialyzed into 0.01M sodium phosphate, 0.07M sodium chloride, pH 7.3, filtered through a 0.22 µm filter, vialed and lyophilized. No preservative added.