Foamtips™ Swabs, ITW Chemtronics
Catalog # 76240-046
Supplier: ITW Chemtronics
Specifications
- Description:Foamtips™ Swabs
- Cat. no.:76240-046
- Packaging:50 swabs per bag
Specifications
About this item
Foamtips™ Swabs are constructed from medical grade foam which features the best open cell structure, providing excellent particle entrapment characteristics. The foam is thermally bonded to the swab handle without using adhesives. These foam swabs are ideal for general cleaning applications.
- Precision cleaning swabs with polyurethane foam head
- Purest medical grade foam
- Traps surface particles while cleaning
- High solvent capacity; holds solvent well
Fiberless construction does not generate loose fibers or particles, no adhesives or binders are used in the construction, economical for high quantity applications. Used for clean excess adhesives after gluing, clean micro-mechanical devices, remove flux residues from printed circuit boards and apply small precise quantities of adhesive or lubricants.