ContactUsLinkComponent
End Cap Clamp, Holland Applied Technologies
Catalog # 76237-894
End Cap Clamp, Holland Applied Technologies
Catalog # 76237-894
Supplier Number:  EG16A-6L.75
Specifications
  • Description:
    End Cap Clamp
  • Material:
    Stainless steel
  • Size:
    3/4"
  • Cat. no.:
    76237-894
Specifications
About this item
Solid end cap is made of stainless steel 304 and clamps on to the end of a process line. The end cap clamps on to the end of a ferruled tube to close the line, providing a non-permanent seal that provides take down access.
This sanitary clamp end cap meets 3-A sanitary standards and is suitable for use in food, beverage, dairy and cosmetic applications.