HY-PRO® NRT® Taps, Sid Tool Co
HY-PRO® NRT® Taps, 0.141" Ø shank, 9/16" thread size, 0.110" (square), 1⅞"
Catalog # 76253-800
Supplier: SID TOOL CO INC TE
Fournisseur #: 9247875
Jump to:
Product Details & Documents
Forming tap with UNC thread.
- Tin coated
- Thread limit H2
- Threads 40 per inch
- Chamfer 2.5 P
Spécifications
- Désignation:HY-PRO® NRT® Taps, 0.141" Ø shank, 9/16" thread size
- Longueur:1⅞"
- Taille:0.110" (square)