QuadraTest Multimode Electronic Test Indicator, Sid Tool | Avantor
QuadraTest Multimode Electronic Test Indicator, Sid Tool
Electronic Test Indicator
Catalog # 76253-182
Supplier:   SID TOOL CO INC TE
Fournisseur #:  20426136
QuadraTest Multimode Electronic Test Indicator, Sid Tool
Catalog # 76253-182
Supplier:  SID TOOL CO INC TE
QuadraTest Multimode Electronic Test Indicator, Sid Tool
Electronic Test Indicator
Price
$6,203.79
Log in pour la disponibilité
Each
Product Details & Documents
Multimode electronic test indicator offers unrivaled measuring versatility in an electronic test indicator!
Available with a 1/2" or 17/16" contact point. The QuadraTest is the perfect solution to test indicating applications.
Features four measuring modes (Minmum, Maximum, TIR and direct), with an RS-232 interface that is water, oil and coolant resistant to IP65 compliancy with or without the interface cable installed! The QuadraTest also features true inch/metric conversion, a 11/2" diameter dial face that rotates a full 360° with both analog bar graph and numeric digital display.
Certifications: Meets or exceeds DIN/ISO standards.
Spécifications
  • Désignation:
    Electronic Test Indicator
  • Cat. No.:
    76253-182
  • Measuring range:
    1/5"/0.5 mm