ContactUsLinkComponent
Corepoint Scientific™ White Diamond Series Refrigerator and Freezer Combination Units with Natural Refrigerants, Horizon Scientific
Corepoint Scientific™ White Diamond Series Refrigerator and Freezer Combination Units with Natural Refrigerants, Horizon Scientific
Supplier:  Horizon Scientific
   Sustainable Options Available
Certificates
undefined
Corepoint Scientific™ White Diamond Series Refrigerator and Freezer Combination Units with Natural Refrigerants, Horizon Scientific
Supplier:  Horizon Scientific
   Sustainable Options Available
Specifications
  • Catalog No:
  • 76300-640
  • 76300-642
  • 76300-644
  • 76300-580
  • 76300-646
  • 76300-582
  • 76300-584
  • 76300-586
  • 76300-636
  • 76300-638
  • 76300-576
  • 76300-578
  • Environmentally Preferable:
  • Safer Human and Environmental Health, Energy Efficient
  • Safer Human and Environmental Health, Energy Efficient
  • Safer Human and Environmental Health, Energy Efficient
  • Safer Human and Environmental Health, Energy Efficient
  • Safer Human and Environmental Health, Energy Efficient
  • Safer Human and Environmental Health, Energy Efficient
  • Safer Human and Environmental Health, Energy Efficient
  • Safer Human and Environmental Health, Energy Efficient
  • Safer Human and Environmental Health, Energy Efficient
  • Safer Human and Environmental Health, Energy Efficient
  • Safer Human and Environmental Health, Energy Efficient
  • Safer Human and Environmental Health, Energy Efficient
  • Cat. No.:
  • 76300-640
  • 76300-642
  • 76300-644
  • 76300-580
  • 76300-646
  • 76300-582
  • 76300-584
  • 76300-586
  • 76300-636
  • 76300-638
  • 76300-576
  • 76300-578
  • Defrost:
  • Auto (Freezer)
    Cycle (Refrigerator)
  • Auto (Freezer)
    Cycle (Refrigerator)
  • Manual (Freezer)
    Cycle (Refrigerator)
  • Manual (Freezer)
    Cycle (Refrigerator)
  • Manual (Freezer)
    Cycle (Refrigerator)
  • Manual (Freezer)
    Cycle (Refrigerator)
  • Manual (Freezer)
    Cycle (Refrigerator)
  • Auto (Freezer)
    Cycle (Refrigerator)
  • Manual (Freezer)
    Cycle (Refrigerator)
  • Manual (Freezer)
    Cycle (Refrigerator)
  • Manual (Freezer)
    Cycle (Refrigerator)
  • Manual (Freezer)
    Cycle (Refrigerator)
  • Shelves/drawers:
  • 6 adjustable shelves (refrigerator), 2 adjustable shelves (freezer)
  • 6 adjustable shelves (refrigerator), 2 adjustable shelves (freezer)
  • 2 adjustable shelves, 1 fixed shelf (refrigerator), 2 fixed shelves (freezer)
  • 2 adjustable shelves, 1 fixed shelf (refrigerator), 2 fixed shelves (freezer)
  • 2 adjustable shelves, 1 fixed shelf (refrigerator), 2 fixed shelves (freezer)
  • 2 adjustable shelves, 1 fixed shelf (refrigerator), 2 fixed shelves (freezer)
  • 3 adjustable shelves (refrigerator), 2 adjustable shelves (freezer)
  • 3 adjustable shelves (refrigerator), 2 adjustable shelves (freezer)
  • 6 adjustable shelves (refrigerator), 2 adjustable shelves (freezer)
  • 6 adjustable shelves (refrigerator), 2 adjustable shelves (freezer)
  • 2 adjustable shelves, 1 fixed hed against back wall (refrigerator); 2 fixed shelves (freezer)
  • 2 adjustable shelves, 1 fixed shelf (refrigerator), 2 fixed shelves (freezer)
  • Door type:
  • 2 solid swing doors, hinge right
  • 2 solid swing doors, hinge right
  • 2 solid swing doors, hinge right
  • 2 stainless steel swing doors, hinge right
  • 2 solid swing doors, hinge left
  • 2 stainless steel swing doors, hinge left
  • 2 exterior swing doors, glass (refrigerator), solid (freezer), hinge right
  • 2 exterior swing doors, glass (refrigerator), solid (freezer), hinge right
  • 2 exterior swing doors, glass (refrigerator), solid (freezer), hinge right
  • 2 solid swing doors, hinge right
  • 2 solid swing doors, hinge right
  • 2 glass swing doors, self-closing, hinge left
  • Electrical:
  • Refrigerator: 115 V, 60 Hz, 2.3 A, 1/3 HP
    Freezer: 115 V, 60 Hz, 1 A, 1/8 HP
  • Refrigerator: 115 V, 60 Hz, 2.3 A, 1/3 HP
    Freezer: 115 V, 60 Hz, 1 A, 1/8 HP
  • Refrigerator: 115 V, 60 Hz, 1.74 A,1/8 HP
    Freezer: 115 V, 60 Hz, 1.5 A, 1/8 HP
  • Refrigerator: 115 V, 60 Hz, 1/8 HP, 1.74 A
    Freezer: 115 V, 60 Hz, 1/8 HP, 1.5 A
  • Refrigerator: 115 V, 60 Hz, 1.74 A,1/8 HP
    Freezer: 115 V, 60 Hz, 1.5 A, 1/8 HP
  • Refrigerator: 115 V, 60 Hz, 1/8 HP, 1.74 A
    Freezer: 115 V, 60 Hz, 1/8 HP, 1.5 A
  • Refrigerator: 115 V, 60 Hz, 1.3 A, 1/6 HP
    Freezer: 115 V, 60 Hz, 1 A, 1/8 HP
  • Refrigerator: 115 V, 60 Hz, 1.3 A, 1/6 HP
    Freezer: 115 V, 60 Hz, 1 A, 1/8 HP
  • Refrigerator: 115 V, 60 Hz, 2.3 A, 1/3 HP
    Freezer: 115 V, 60 Hz, 1 A, 1/8 HP
  • Refrigerator: 115 V, 60 Hz, 2.3 A, 1/3 HP
    Freezer: 115 V, 60 Hz, 1 A, 1/8 HP
  • Refrigerator: 115 V, 60 Hz, 1/8 HP, 1.74 A
    Freezer: 115 V, 60 Hz, 1/8 HP, 1.5 A
  • Refrigerator: 115 V, 60 Hz, 1/8 HP, 1.74 A
    Freezer: 115 V, 60 Hz, 1/8 HP, 1.5 A
  • Weight:
  • 131.1 kg (289 lbs.)
  • 131.1 kg (289 lbs.)
  • 122 kg (269 lbs.)
  • 120.7 kg (266 lbs.)
  • 122 kg (269 lbs.)
  • 120.7 kg (266 lbs.)
  • 93.9 kg (207 lbs.)
  • 93.9 kg (207 lbs.)
  • 131.1 kg (289 lbs.)
  • 131.1 kg (289 lbs.)
  • 127.9 kg (282 lbs.)
  • 127.9 kg (282 lbs.)
  • Environmental certification:
  • HFC Free
  • HFC Free
  • HFC Free
  • HFC Free
  • HFC Free
  • HFC Free
  • HFC Free
  • HFC Free
  • HFC Free
  • HFC Free
  • HFC Free
  • HFC Free