ESD Kit, Production Basics
ESD kit
Catalog # 76248-536
Supplier: Production Basics
Fournisseur #: PB-8991
Jump to:
Product Details & Documents
Kit includes a tapered ground bolt for ESD work bench and a 6' ground cord.
Spécifications
- Désignation:ESD kit