Slide Switch, Digi-Key
Silver, Copper, –10...104 °C, 125 V
Catalog # 76250-376
Supplier: DIGI-KEY CORPORATION
Supplier #: GF-126-3011
Jump to:
Product Details & Documents
Miniature panel mount and through hole slide switch.
- Vertical orientation
- Two through five positions
- Detent or spring return
Specifications
- Tension:125 V
- Color:Silver
- Matière:Copper
- Operating Temperature:–10...104 °C