Peg Board/Bin Panel Enclosure, Production Basics
76248-512
- Width:72"
- Color:Black
- Height:20"
- Cat. no.:76248-512
- Weight:30 lbs.
Using two of the hardest working Enclosures to organize tools and supplies, the peg board accepts standard pegs for hanging tools and the bin panel side accepts standard storage bins.
- Ideal for assembly-intensive work spaces that require high-volume, high-mix supplies
- Full multi-panel is finished in standard black wrinkle epoxy powder coat
This multi-panel is shipped assembled, but sides can be easily switched depending on operator preference.