ACT Cable Tie, Testequity LLC TE
UV Black Cable Ties, Black, 11.25", 0.180", 3.062"
Catalog # 76248-090
Supplier: TESTEQUITY LLC TE
Fournisseur #: AL-11-50-0-C
Jump to:
Product Details & Documents
Miniature Cable Ties, except Heat Stabilized Black Cable Ties, are Mil-Spec Approved.
Certifications: ROHS Compliant.
Spécifications
- Ø:3.062"
- Largeur:0.180"
- Color:Black
- Désignation:UV Black Cable Ties
- Longueur:11.25"