Lead Bender Forming Tool, Testequity LLC TE
51/4", ABS plastic, high impact, 1 Watt Resistors and 2 Watt Resistors, 0.800 - 1.900"
Catalog # 76252-996
Supplier: TESTEQUITY LLC TE
: 119-669
Jump to:
Product Details & Documents
Radiuses edges
Spécifications
- Matière:ABS plastic, high impact
- Taille:51/4"
- Type:1 Watt Resistors and 2 Watt Resistors
- Width:0.800 - 1.900"