Wire, Cable Labels, Brady
Wire marking sleeve 1.5", matte finish, die-cut, Cloth, 3, Black on white, 0.0088", 25 cards/pack
Catalog # 76250-238
Supplier: Brady Worldwide
Fournisseur #: 112225
Jump to:
Product Details & Documents
These B-500 repositionable vinyl cloth wire markers are perfect for general wire marking needs.
- Card-mounted wire markers allow for quick, on-the-spot wire marking
- Excellent flexibility for wrapping around curved surfaces
- Withstands moderate heat, oil, water and dirt
The vinyl cloth holds tightly to surfaces but allows item to be repositioned with ease if necessary.
Abrasion, corrosion, humidity, oil, UV, water and weather-resistant.
Spécifications
- Color:Black on white
- Désignation:Wire marking sleeve 1.5", matte finish, die-cut
- Matière:Cloth
- Épaisseur:0.0088"
- Packaging:25 cards/pack
- Legend:3