Bottoming Chamfer, Sid Tool
Bottoming Chamfer, Sid Tool
Bottoming Chamfer, Sid Tool
Bottoming chamfers are used for threading blind holes to the bottom. They have 1 to 2 threads. Bright provides a smooth, polished finish on the tool. It increases chip flow in softer materials such as aluminum, wood and plastic.
High Speed Steel (HSS) provides good wear-resistance and can be used in general purpose applications for both ferrous and non-ferrous materials.
Product Family Options
  • Désignation
    Matière
    Longueur
    Finish
    Shank Diameter
    Thread Length
  • Bottoming Chamfer Three Flutes, Heli-Coil
    High Speed Steel
    2⅜"
    Bright
    0.2200"
    ¹⁵⁄₁₆"
    Each
    Each - $193.72
    $193.72
    Each
       $193.72
     : 64832033
  • Bottoming Chamfer, Balax
    High Speed Steel
    2½"
    Bright
    0.2500"
    1"
    Each
    Each - $176.29
    $176.29
    Each
       $176.29
     : 64047673