Xcelite® Pro-Series Electronic Screwdrivers, Apex
Catalog # 76244-380
Supplier: Cooper Tools
Specifications
- Cat. no.:76244-380
- Packaging:Boxed
- Blade Length:102 mm (4")
- Blade Diameter:3.97 mm (⁵⁄₃₂")
- Blade Type:Slotted
Specifications
About this item
These screwdrivers feature hard shockproof plastic handle core for durability, while the outer coating is soft rubber for comfort and slip resistance.
- ESD safe hard plastic core to protect sensitive components
- Narrow pinch grip for comfort and control
- Thumb bolster allows added downward force
- Satin finish chrome molybdenum steel shafts provide durability
- Specially treated black oxide tips eliminate light reflection
The handles are ergonomically designed, allowing sustained high torque with minimal slippage and muscle fatigue. Designed with smooth shatterproof seamless handle cap for minimal friction between hand and handle when screwdriver is being turned.