Eclipse Electronics Tool Kit, Global Industrial | Avantor
Eclipse Electronics Tool Kit, Global Industrial
Eclipse Electronics Tool Kit
Catalog # 76250-702
Fournisseur #:  WGB1930009
Eclipse Electronics Tool Kit, Global Industrial
Catalog # 76250-702
Eclipse Electronics Tool Kit, Global Industrial
Eclipse Electronics Tool Kit
Price
$1,728.22
Log in pour la disponibilité
Each Kit of 50
Product Details & Documents
 : Kit includes long nose plier (135 mm), diagonal cutting plier (110 mm), dual color lineman's plier (210 mm), dual color long nose plier (165 mm), adjustable wrench (6"), dual color side cutting plier (165 mm), needle file set (5 pieces ), ceramic soldering iron (110 V), 31/2 compact digital multimeter, measuring tape (3 m/10 ft.), solder core (63%), SN, hex key set (7 pieces) , wire stripper cutter, utility knife (3 blades self loading), all purpose snip, solder aid tools, desoldering pump, electronic combination wrench (10 pieces), flux bottle (2 pieces), blow brush (Ø 62 mm), IC extractor, slip joint plier (154 mm), pro-soft screwdriver (3.0×75 mm, #0×75 mm, 5.0×75 mm, #1×75 mm, 6.0×100 mm, #2×100 mm, #2×250 mm, 6.0×40 mm, #2×40 mm, 2.0×50 mm, 2.4×75 mm, 3.0×100 mm, #00×50 mm, #0×75 mm, #1×100 mm, soldering tip cleaner, fine tip straight tweezer, utility component storage box, hobby vise, top pallet, aluminum frame tool case with one pallet.
Spécifications
  • Désignation:
    Eclipse Electronics Tool Kit