Chamfer Tools, Cutting Tool Supply
MCH-2R Single End and MCH-2D Double End Chamfer Tool, with 2 Flutes, Steel, 3", ½", 82°, ½"
Catalog # 76249-864
Supplier: CUTTING TOOL SUPPLY TE
Fournisseur #: B10226
Jump to:
Product Details & Documents
Chamfer tools made of solid carbide.
Suitable for cutting steel, cast iron and non-ferrous materials.
Spécifications
- Désignation:MCH-2R Single End and MCH-2D Double End Chamfer Tool, with 2 Flutes
- Longueur:3"
- Matière:Steel
- Shank Diameter:½"
- Cutting Diameter:½"
- Ratcheting Angle:82°