Kilo Scale Add-On Distillation Kit, Ace Glass Incorporated
Catalogue # 76291-814
Fournisseur: Ace Glass
Spécifications
- Description:Kilo-Scale Add-On Distillation Kit
- Environmentally Preferable:
- Cat. no.:76291-814
Spécifications
À propos de cet article
Add-on distillation kit for use with the ACE Universal Stand as part of any ACE kilo scale reactor. Kit includes a side shelf to attach to an existing stand, a receiving flask, a distillation splitter, a condenser, tubing, and adapters.