Stem Casters, Quantum Storage Systems
Stem casters
  76250-714
 :  WR-00H-B
Stem Casters, Quantum Storage Systems
  76250-714
Stem Casters, Quantum Storage Systems
Stem casters
$539.94
Each
The stem caster comes with four swivel 5×11/4" polyurethane casters.
  • Two casters have brakes
  • Each caster is capable of carrying up to 300 lbs.
  • Can be attached to a variety of shelving units to make them mobile and easy to move around
  • Can also be used to transport stored parts
Each caster has been constructed to handle heavy loads.
  • Désignation:
    Stem casters