Pillowstat ESD Boxes, Conductive Containers
Supplier: CONDUCTIVE CONTAINERS INC. TE
Jump to:
À propos de cet article
Super soft ESD foam for surface mount technology.
- ESD safe foam forms around the parts
- No tooling required
- Provides great physical protection
- Fits multiple sized components
Ideal for fragile, sensitive and expensive components.
3 Options Available Below
Product Details & Documents
Super soft ESD foam for surface mount technology.
- ESD safe foam forms around the parts
- No tooling required
- Provides great physical protection
- Fits multiple sized components
Ideal for fragile, sensitive and expensive components.
Product Family Options
Informations sur le produit
- DésignationDimensionsAvailabilityPrice
- Supplier Number: PP1010
- Supplier Number: 18M-1304-R1
- Supplier Number: PP3545