Avantik boxed loose standard tissue processing/embedding cassettes with lid are made with the highest quality and provide excellent value.
- Anterior writing area is at a 35° angle
- Three-sided writing surface
Molded from a high density polymer, these cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. The snap latch and hinge-lock design prevents early separation of base and lid, and allows for one-handed operation. Lids can be opened and closed as often as necessary, and they always relock securely without danger of specimen loss.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.