To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Pack type:Square polypropylene bottle inside carton
- Particle Size:35 µm
- Pore size:500 Å
- Description:BAKERBOND® XWP 500 PolyCSX-35
- Size:500 ml
- Environmentally Preferable:
- Cat. no.:CAJT7587-03
The J.T.Baker® BAKERBOND® PolyCSX multimode ion exchange (IEX) chromatography resin functions as a strong cation exchange resin with improved selectivity for superior separation in process chromatography applications.
Powered by a proprietary surface chemistry combining strong cation exchange functionality with weak cationic and anionic exchange sites, BAKERBOND® PolyCSX multimode cation exchange chromatography media utilizes its unique selectivity to enable higher purification performance across a wide range of biopharmaceuticals.
Designed and manufactured by Avantor to the high standards established for our J.T.Baker® brand chemicals, BAKERBOND® PolyCSX ion exchange resin can be used with other J.T.Baker® BAKERBOND® resins to enhance the separation power of your chromatography process steps.