Hinged Conductive Boxes, All-Spec Industries
Supplier:   HISCO, INC TE
Hinged Conductive Boxes, All-Spec Industries
Supplier:  HISCO, INC TE
Certificates
Hinged Conductive Boxes, All-Spec Industries
2 Options Available Below
Product Details & Documents
These 660 series conductive plastic boxes are made of a surface and volume conductive materials and it also a volume conductive and electrostatic shielding material.
 : Accordance to EIA standard 514, ASTM D257-75 and EOS/ESD-DS-11.11.1991 test methods.
Product Family Options
Informations sur le produit
  • Color
    Matière
    No. of Compartments
    Dimensions
    Availability
    Price
  • Black
    Plastic
    1
    1×1×1" (Compartments)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $2,839.18
    $2,839.18
    Log in pour la disponibilité
    Each
    Each - $2,839.18
    $2,839.18
    Case of 1,000
       $2,839.18
    Each
       $2,839.18
    Supplier Number: 663-111
  • Black
    Plastic
    1
    7×5×13/4" (Compartments)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $58.21
    $58.21
    Each
       $58.21
    Supplier Number: 665-752