To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen symbol:ACSS2
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Western blot:Yes
- Size:100 µg
- Storage temperature:Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
- Shipping temperature:Dry Ice
- Purification:Affinity purified
- Cat. no.:76262-958
This antibody is designed, produced, and validated as part of a collaboration between Rockland and the National Cancer Institute (NCI). ACSS2 (Acyl-CoA synthetase short-chain family member 2) is a protein coding gene encoding a cytosolic enzyme that catalyzes the activation of acetate for use in lipid synthesis and energy generation. This protein acts as a monomer and produces acetyl-CoA from acetate in a reaction that requires ATP. Expression of this gene is regulated by sterol regulatory element-binding proteins, transcription factors that activate genes required for the synthesis of cholesterol and unsaturated fatty acids. This protein is upregulated in many cancers and is important for pro-survival under hypoxic conditions. ACSS2 may be associated with disorders such as Cystoisosporiasis. Anti-ACSS2 Antibody is useful for researchers interested in parasites, intestines, Glucose Energy Metabolism, and AKT research.
Anti-ACSS2 Antibody has been tested in Western Blot and ELISA. Expect a band at approximately 76 kDa in western blot using appropriate lysates. Positive control whole cell lysates used Hep-G2 at 1µg/mL for WB. Although not tested, this antibody is likely functional in immunohistochemistry, immunofluorescence, and immunoprecipitation.
Type: Primary
Antigen: ACSS2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: