Single Flute Countersink, Cutting Tool Supply | Avantor
Single Flute Countersink, Cutting Tool Supply
Single Flute Countersink, Cutting Tool Supply
Single Flute Countersink, Cutting Tool Supply
Made of black oxide with reduced shank.
Certifications: ASME B94.11M.
Product Family Options
  • Désignation
    Diameter
    Longueur
    Shank Diameter
  • Single Flute Countersink, 100°
    5/8"
    2¼"
    ⅝"
    Each
    Each - $189.94
    $189.94
    Each
       $189.94
     : C46122
  • Single Flute Countersink, 90°
    1"
    2¾"
    ½"
    Each
    Each - $419.66
    $419.66
    Each
       $419.66
     : C46132
  • Single Flute Countersink, 100°
    1"
    2¾"
    ½"
    Each
    Each - $426.73
    $426.73
    Each
       $426.73
     : C46133
  • Single Flute Countersink, 100°
    3/4"
    2⅝"
    ½"
    Each
    Each - $247.55
    $247.55
    Each
       $247.55
     : C46127