Tester, R-C-L Meter, Excelta Corp® | Avantor
Tester, R-C-L Meter, Excelta Corp®
RCL meter, straight, 5 star quality, length 100 mm (3.94"), tip size 0.5×0.25 mm (0.02×0.01"), hand held, bluetooth
Catalog # 76471-438
Supplier:   Excelta
Fournisseur #:  TM-500-BTW
Tester, R-C-L Meter, Excelta Corp®
Catalog # 76471-438
Supplier:  Excelta
Tester, R-C-L Meter, Excelta Corp®
Tester, R-C-L Meter, Excelta Corp®
RCL meter, straight, 5 star quality, length 100 mm (3.94"), tip size 0.5×0.25 mm (0.02×0.01"), hand held, bluetooth
Price
$1,368.90
Log in pour la disponibilité
Each
Product Details & Documents
The TM-500 five star IntelliTweeze™ is a quick and easy way of testing and troubleshooting many types of components.
  • Easy way of testing and troubleshooting many types of components
It features built-in high precision LCR probe, 0.2 to 0.5% base measurement accuracy, automated component identification, automated test range selectioon, manual C,R,L,Z and ESR modes, adjustable test signal levels, diade polarity short testing, secondary D,Q, and ESR parameters, high visibility LED display and build-in Li-Ion battery.
Spécifications
  • Désignation:
    RCL meter, straight, 5 star quality, length 100 mm (3.94"), tip size 0.5×0.25 mm (0.02×0.01"), hand held, bluetooth