ContactUsLinkComponent
Conductive Plugs, CEP Series, Caplugs
Catalogue # 76244-284
Fournisseur:  Caplugs
Conductive Plugs, CEP Series, Caplugs
Catalogue # 76244-284
Fournisseur:  Caplugs
Supplier Number:  CEP-10
Spécifications
  • Color:
    Black
  • Description:
    Connective Plugs, Ø 19.30 mm
  • Height:
    14.22 mm
  • Cat. no.:
    76244-284
Spécifications
À propos de cet article
Designed to plug the more popular sizes of both threaded and non-threaded, bayonet type electrical and electronic connectors, CEP Conductive Plugs provide electrical shielding as well as mechanical protection.
  • Surface resistivity of less than 1×105 ohms