Tool Extractor, Jensen Tools
PLCC Extractor
Catalog # 76245-298
Supplier: TESTEQUITY LLC TE
Supplier #: EX5
Jump to:
Product Details & Documents
These extracter use to rmeove the chip, range of PLCC sizes from 20 pins through 84 pins from any socket without damage to the chip.
- ESD safe handles for use on all static safe components
- Made of stainless steel
Spring loaded one-hand design requires no pulling, just squeeze handles and chip is lifted from the socket.
Specifications
- Désignation:PLCC Extractor