10X Tris-Borate-EDTA (TBE) Buffer | Avantor
10X Tris-Borate-EDTA (TBE) Buffer
10X Tris-Borate-EDTA TBE Buffer, 4 L
Catalog # 76236-396
Fournisseur #:  MB-019-4000
10X Tris-Borate-EDTA (TBE) Buffer
Catalog # 76236-396
CAS Number:  
10X Tris-Borate-EDTA (TBE) Buffer
10X Tris-Borate-EDTA TBE Buffer, 4 L
CAS Number:  
Price
$339.84
Log in pour la disponibilité
Each (4l)
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
TBE is often used in procedures involving nucleic acids, the most common being electrophoresis in molecular biology.
  • Concentrated 10X
  • 0.9 M Tris-Borate, 0.01 M EDTA at a pH of 8.3
Tris-acid solutions are effective buffers for slightly basic conditions, which keep DNA deprotonated and soluble in water. EDTA is a chelator of divalent cations, particularly of magnesium (Mg2+). As these ions are necessary co-factors for many enzymes, including contaminant nucleases, the role of the EDTA is to protect the nucleic acids against enzymatic degradation. But since Mg2+ is also a co-factor for many useful DNA-modifying enzymes such as restriction enzymes and DNA polymerases.
Application Note: This product is a concentrated stock solution and should be diluted appropriately with distilled, deionized water or equivalent to its final working concentration. 10X Tris-Borate EDTA (TBE) consists of 0.9 M Tris-Borate, 0.01 M EDTA at a pH of 8.3.  Meticulously prepared using ultra pure reagents dissolved in highly polished pharmaceutical grade deionized water.
Spécifications
Spécifications
  • Désignation:
    10X Tris-Borate-EDTA TBE Buffer
  • Taille:
    4 L