ESD Jacket, Snap Cuff, Transforming Technologies
Supplier: Transforming Technologies
À propos de cet article
These collared jackets are made fromlightweight 90% polyester and 10% carbon fabric for supreme comfort and ESD shielding.
Features a snap front closure, wrist adjustment snaps and three roomy patch pockets. Maintains consistent continuity readings even after 100 wash cycles.
: Meets or exceeds ANSI ESD-S20.20 requirements.
3 Options Available Below
Product Details & Documents
These collared jackets are made fromlightweight 90% polyester and 10% carbon fabric for supreme comfort and ESD shielding.
Features a snap front closure, wrist adjustment snaps and three roomy patch pockets. Maintains consistent continuity readings even after 100 wash cycles.
: Meets or exceeds ANSI ESD-S20.20 requirements.
Specifications for Product Family
Spécifications
- Catalog No:
- 76249-818
- 76249-418
- 76249-820
- Garment application:
- General Purpose
- Industrial|General Purpose
- General Purpose
- Risk area:
- ESD
- ESD
- ESD
- Gender:
- Unisex
- Unisex
- Unisex
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- 90% polyester, 10% carbon fabric
- 90% polyester, 10% carbon fabric
- 90% polyester, 10% carbon fabric
Spécifications
Product Family Options
Informations sur le produit
- ColorTaille de confectionAvailabilityPrice
- Supplier Number: JKC9024SPBK-LSpécifications:
More
Specifications
Cat. No.76249-818Garment applicationGeneral PurposeRisk areaESDGenderUnisexClosure typePress studs / snapsFabric90% polyester, 10% carbon fabric - Supplier Number: JKC9025SPBK-XLSpécifications:
More
Specifications
Cat. No.76249-418Garment applicationIndustrial|General PurposeRisk areaESDGenderUnisexClosure typePress studs / snapsFabric90% polyester, 10% carbon fabric - Supplier Number: JKC9026SPBK-XXSpécifications:
More
Specifications
Cat. No.76249-820Garment applicationGeneral PurposeRisk areaESDGenderUnisexClosure typePress studs / snapsFabric90% polyester, 10% carbon fabric