J.T.Baker® BAKERBOND®, PolyABx Multimode Ion Exchange Chromatography Resin
J.T.Baker® BAKERBOND®, PolyABx Multimode Ion Exchange Chromatography Resin
MOC:  Non réglementé, avec information
Certificates
J.T.Baker® BAKERBOND®, PolyABx Multimode Ion Exchange Chromatography Resin
J.T.Baker® BAKERBOND®, PolyABx Multimode Ion Exchange Chromatography Resin
MOC:  Non réglementé, avec information
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The J.T.Baker® BAKERBOND® PolyABx multimode ion exchange (IEX) chromatography resin functions as a weak cation exchange resin with improved selectivity for superior separation in process chromatography applications.
Powered by a proprietary surface chemistry combining weak cation exchange functionality with weak anionic exchange sites and hydrophilicity, BAKERBOND® PolyABx multimode cation exchange chromatography media utilizes its unique selectivity to enable higher purification performance across a wide range of biopharmaceuticals.
Designed and manufactured by Avantor to the high standards established for our J.T.Baker® brand chemicals, BAKERBOND® PolyABx ion exchange resin can be used with other J.T.Baker® BAKERBOND® resins to enhance the separation power of your chromatography process steps.
Documents
Product Family Options
Informations sur le produit
  • Désignation
    Granulométrie
    Taille
    Taille de pore
    Type de conditionnement
    Availability
    Price
  • PolyABx, primarily weak cation exchange media
    35 µm
    100 ml
    500 Å
    Square natural PP bottle
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (100ml)
    Each (100ml) - $628.19
    $628.19
    Each (100ml)
       $628.19
    Supplier Number: BAKR7586-02
    Spécifications:
    MOC:  Non réglementé, avec information
    MOC:  Non réglementé, avec information
  • PolyABx, primarily weak cation exchange media
    35 µm
    5 L
    500 Å
    Polypropylene bottle inside poly PA
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (5l)
    Each (5l)
    Supplier Number: BAKR7586-05
    Spécifications:
    MOC:  Non réglementé, avec information
    MOC:  Non réglementé, avec information