Kimble® Potter-Elvehjem Tissue Grinder Components, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
A graduated glass tube and a finely machined PTFE pestle comprise this popular style.
- Graduations are fused into the glass
- Replacement components are completely interchangeable
Radial serrations on the lower portion of the pestle deliver the homogenate into the cylindrical portion of the tube.
Frequently Bought Together