Order Entry
Canada
ContactUsLinkComponent
Rat KIM1 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-658
CAS Number:  
Rat KIM1 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-658
Supplier Number:  KOA0531
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Description:
    Rat KIM1 ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76235-658

Specifications

About this item

Rat KIM1 AccuSignal ELISA Kit

  • Natural and recombinant rat KIM1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: S18-V238

KIM1(TIM-1), also known as Hepatitis A virus cellular receptor 1, is a protein that in Rat is encoded by the HAVCR1 gene. Infection of canine osteogenic sarcoma cells expressing HAVCR1 with HAV led to conclude that the protein is indeed a receptor for the virus. Immunofluorescence microscopy demonstrated internalization of HAV by dog cells expressing HAVCR1. Using a monoclonal antibody to rat Tim1, Tim1 was expressed after activation of naive T cells and on T cells differentiated in Th2-polarizing conditions. By homology of synteny with the rat Tim1 gene and database analysis, was mapped the HAVCR1 gene to 5q33.2.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml rat KIM1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, tissue homogenates, serum, plasma(heparin, EDTA) or urine to each empty well. It is recommended that each rat KIM1 standard solution and each sample be measured in duplicate.