Red Blood Cell Depletion Magnetic Beads
- Separation of Human Red Blood Cells, from whole blood.
- Remaining white blood cells can be used for downstream assays such as immunoassays, Western blots, FACS, and immunohistochemistry.
- Bead mean diameter is ~0.5 μm, bead concentration is 20 mg/mL.
Red Blood Cell Depletion Magnetic Beads recognizes human red blood cells. Red Blood Cell Depletion Magnetic Beads is recommended for the depletion of human Red Blood Cells in samples prior to downstream immunological and non-immunological assays and has been tested in Flow Cytometry. Some additional assays could include Colony Forming Unit (CFU) Assay and Immunohistochemistry among others. These magnetic beads are uniform, non-aggregating, super-paramagnetic beads consisting of a ferric oxide core functionalized with various silane groups. The beads have a large surface area with high capture efficiencies.
Red Blood Cell Depletion Magnetic Beads can be used for separation of Red Blood Cells, from whole blood. Remaining white blood cells can be used for downstream assays such as immunoassays, Western blots, FACS, and immunohistochemistry. For 100 µL of whole blood cells add 75 µL of Depletion beads, expecting 75 – 95% depletion of RBC after one pass. Optimal amount for applications should be determined by the researcher.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 76221-886
- 76221-884
- Shipping conditions:
- Wet Ice
- Wet Ice